Assay Details
| Gene Symbol: slc4a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034984.1 | NP_001030156.1 | 2 - 3 | 104 | 86 | |
| XM_005165244.4 | XP_005165301.1 | 2 - 3 | 381 | 86 | ||
| XM_009301742.3 | XP_009300017.1 | 1 - 2 | 312 | 86 | ||
| XM_009301743.3 | XP_009300018.1 | 1 - 2 | 311 | 86 | ||
| XM_009301747.3 | XP_009300022.1 | 1 - 2 | 311 | 86 | ||
| XM_021476291.1 | XP_021331966.1 | 3 - 4 | 301 | 86 | ||
| XM_021476293.1 | XP_021331968.1 | 3 - 4 | 274 | 86 | ||
| XM_021476294.1 | XP_021331969.1 | 3 - 4 | 243 | 86 | ||
| GenBank mRNA | AY727858.1 | - | 2 - 3 | 104 | 86 | |
| BC162559.1 | - | 2 - 3 | 163 | 86 | ||
More Information
Set Membership: |
|

slc4a4a
Genomic Map