Assay Details
| Gene Symbol: slc4a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037237.1 | NP_001032314.1 | 19 - 20 | 3243 | 76 | |
| XM_005163511.4 | XP_005163568.1 | 21 - 22 | 3428 | 76 | ||
| XM_009298626.3 | XP_009296901.1 | 20 - 21 | 3403 | 76 | ||
| XM_009298629.3 | XP_009296904.1 | 19 - 20 | 3346 | 76 | ||
| XM_021480957.1 | XP_021336632.1 | 21 - 22 | 3368 | 76 | ||
| XM_021480959.1 | XP_021336634.1 | 20 - 21 | 3302 | 76 | ||
| GenBank mRNA | AY876015.1 | - | 19 - 20 | 3243 | 76 | |
| DQ005500.1 | - | 5 - 6 | 789 | 76 | ||
More Information
Set Membership: |
|

slc4a2a
Genomic Map