Assay Details
| Gene Symbol: slc4a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037237.1 | NP_001032314.1 | 20 - 21 | 3510 | 60 | |
| XM_005163511.4 | XP_005163568.1 | 22 - 23 | 3695 | 60 | ||
| XM_009298626.3 | XP_009296901.1 | 21 - 22 | 3670 | 60 | ||
| XM_009298629.3 | XP_009296904.1 | 20 - 21 | 3613 | 60 | ||
| XM_021480957.1 | XP_021336632.1 | 22 - 23 | 3635 | 60 | ||
| XM_021480959.1 | XP_021336634.1 | 21 - 22 | 3569 | 60 | ||
| GenBank mRNA | AY876015.1 | - | 20 - 21 | 3510 | 60 | |
| DQ005500.1 | - | 6 - 7 | 1056 | 60 | ||
| DQ005501.1 | - | 5 - 6 | 852 | 60 | ||
More Information
Set Membership: |
|

slc4a2a
Genomic Map