Assay Details
| Gene Symbol: slc4a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037237.1 | NP_001032314.1 | 22 - 23 | 3851 | 120 | |
| XM_005163511.4 | XP_005163568.1 | 24 - 25 | 4036 | 120 | ||
| XM_009298626.3 | XP_009296901.1 | 23 - 24 | 4011 | 120 | ||
| XM_009298629.3 | XP_009296904.1 | 22 - 23 | 3954 | 120 | ||
| XM_021480957.1 | XP_021336632.1 | 24 - 25 | 3976 | 120 | ||
| XM_021480959.1 | XP_021336634.1 | 23 - 24 | 3910 | 120 | ||
| GenBank mRNA | AY876015.1 | - | 22 - 23 | 3851 | 120 | |
| DQ005500.1 | - | 8 - 9 | 1397 | 120 | ||
More Information
Set Membership: |
|

slc4a2a
Genomic Map