Assay Details
| Gene Symbol: slc4a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037237.1 | NP_001032314.1 | 4 - 5 | 715 | 96 | |
| XM_005163511.4 | XP_005163568.1 | 5 - 6 | 843 | 96 | ||
| XM_009298626.3 | XP_009296901.1 | 4 - 5 | 818 | 96 | ||
| XM_009298629.3 | XP_009296904.1 | 4 - 5 | 818 | 96 | ||
| XM_021480957.1 | XP_021336632.1 | 5 - 6 | 783 | 96 | ||
| XM_021480959.1 | XP_021336634.1 | 4 - 5 | 717 | 96 | ||
| GenBank mRNA | AY876015.1 | - | 4 - 5 | 715 | 96 | |
More Information
Set Membership: |
|

slc4a2a
Genomic Map