Assay Details
| Gene Symbol: borcs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039988.1 | NP_001035077.1 | 3 - 4 | 251 | 122 | |
| GenBank mRNA | BC097219.1 | - | 3 - 4 | 251 | 122 | |
More Information
Set Membership: |
|

borcs7
Genomic Map