Assay Details
| Gene Symbol: borcs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039988.1 | NP_001035077.1 | 4 - 5 | 268 | 66 | |
| GenBank mRNA | BC097219.1 | - | 4 - 5 | 268 | 66 | |
More Information
Set Membership: |
|

borcs7
Genomic Map