Assay Details
| Gene Symbol: ormdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040048.1 | NP_001035137.1 | 1 - 2 | 188 | 64 | |
| GenBank mRNA | BC114303.1 | - | 1 - 2 | 188 | 64 | |
More Information
Set Membership: |
|

ormdl2
Genomic Map