Assay Details
| Gene Symbol: ormdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040048.1 | NP_001035137.1 | 2 - 3 | 339 | 79 | |
| GenBank mRNA | BC114303.1 | - | 2 - 3 | 339 | 79 | |
More Information
Set Membership: |
|

ormdl2
Genomic Map