Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 22 - 23 | 2974 | 89 | |
| XM_005171778.4 | XP_005171835.1 | 23 - 24 | 3149 | 89 | ||
| XM_005171779.4 | XP_005171836.1 | 23 - 24 | 3146 | 89 | ||
| XM_021475966.1 | XP_021331641.1 | 21 - 22 | 2942 | 89 | ||
| GenBank mRNA | BC115049.1 | - | 10 - 11 | 1057 | 89 | |
| BC133980.1 | - | 7 - 8 | 822 | 89 | ||
| BC171681.1 | - | 22 - 23 | 3008 | 89 | ||
More Information
Set Membership: |
|

ehmt1a
Genomic Map