Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 16 - 17 | 2301 | 74 | |
| XM_005171778.4 | XP_005171835.1 | 17 - 18 | 2473 | 74 | ||
| XM_005171779.4 | XP_005171836.1 | 17 - 18 | 2470 | 74 | ||
| XM_021475966.1 | XP_021331641.1 | 15 - 16 | 2266 | 74 | ||
| GenBank mRNA | BC115049.1 | - | 4 - 5 | 381 | 74 | |
| BC171681.1 | - | 16 - 17 | 2332 | 74 | ||
More Information
Set Membership: |
|

ehmt1a
Genomic Map