Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 18 - 19 | 2518 | 66 | |
| XM_005171778.4 | XP_005171835.1 | 19 - 20 | 2690 | 66 | ||
| XM_005171779.4 | XP_005171836.1 | 19 - 20 | 2687 | 66 | ||
| XM_021475966.1 | XP_021331641.1 | 17 - 18 | 2483 | 66 | ||
| GenBank mRNA | BC115049.1 | - | 6 - 7 | 598 | 66 | |
| BC133980.1 | - | 3 - 4 | 363 | 66 | ||
| BC171681.1 | - | 18 - 19 | 2549 | 66 | ||
More Information
Set Membership: |
|

ehmt1a
Genomic Map