Assay Details
| Gene Symbol: cirbpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040321.2 | NP_001035411.2 | 6 - 7 | 743 | 87 | |
| NR_132113.1 | 6 - 7 | 750 | 87 | |||
| GenBank mRNA | BC048027.1 | - | 6 - 7 | 689 | 87 | |
| BC115177.1 | - | 5 - 6 | 640 | 88 | ||
| EH436305.1 | - | 6 - 7 | 706 | 87 | ||
| EH507728.1 | - | 6 - 7 | 718 | 87 | ||
More Information
Set Membership: |
|

cirbpb
Genomic Map