Assay Details
| Gene Symbol: pitx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040346.2 | NP_001035436.3 | 1 - 2 | 259 | 100 | |
| XM_021468902.1 | XP_021324577.1 | 2 - 3 | 220 | 100 | ||
| GenBank mRNA | BC115352.1 | - | 1 - 2 | 211 | 100 | |
| EU032338.1 | - | 1 - 2 | 259 | 100 | ||
| EU580407.1 | - | 1 - 2 | 306 | 100 | ||
More Information
Set Membership: |
|

pitx1
Genomic Map