Assay Details
| Gene Symbol: tnfsf10l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042713.1 | NP_001036178.1 | 3 - 4 | 320 | 94 | |
| GenBank mRNA | BC162834.1 | - | 3 - 4 | 402 | 94 | |
| BC162841.1 | - | 3 - 4 | 402 | 94 | ||
| DQ812113.1 | - | 3 - 4 | 320 | 94 | ||
More Information
Set Membership: |
|

tnfsf10l3
Genomic Map