Assay Details
| Gene Symbol: ptpn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044313.2 | NP_001037778.2 | 25 - 26 | 2694 | 102 | |
| XM_005173946.4 | XP_005174003.1 | 26 - 27 | 3105 | 102 | ||
| XM_009296096.3 | XP_009294371.1 | 26 - 27 | 3102 | 102 | ||
More Information
Set Membership: |
|

ptpn4b
Genomic Map