Assay Details
| Gene Symbol: ptpn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044313.2 | NP_001037778.2 | 9 - 10 | 766 | 105 | |
| XM_005173946.4 | XP_005174003.1 | 10 - 11 | 1162 | 105 | ||
| XM_009296096.3 | XP_009294371.1 | 10 - 11 | 1162 | 105 | ||
More Information
Set Membership: |
|

ptpn4b
Genomic Map