Assay Details
| Gene Symbol: prkchb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044806.1 | NP_001038271.1 | 5 - 6 | 673 | 73 | |
| XM_005160500.4 | XP_005160557.1 | 5 - 6 | 898 | 73 | ||
| GenBank mRNA | BC163148.1 | - | 5 - 6 | 771 | 73 | |
More Information
Set Membership: |
|

prkchb
Genomic Map