Assay Details
| Gene Symbol: zpax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044869.3 | NP_001038334.3 | 2 - 3 | 206 | 86 | |
| GenBank mRNA | BC139580.1 | - | 2 - 3 | 206 | 86 | |
More Information
Set Membership: |
|

zpax2
Genomic Map