Assay Details
| Gene Symbol: slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045020.1 | NP_001038485.1 | 1 - 2 | 154 | 77 | |
| XM_009294893.3 | XP_009293168.1 | 2 - 3 | 569 | 77 | ||
| GenBank mRNA | BC163234.1 | - | 1 - 2 | 190 | 77 | |
More Information
Set Membership: |
|

slc30a2
Genomic Map