Assay Details
| Gene Symbol: car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045139.1 | NP_001038604.1 | 6 - 7 | 617 | 102 | |
| XM_005169221.4 | XP_005169278.1 | 6 - 7 | 1095 | 102 | ||
More Information
Set Membership: |
|

car15
Genomic Map