Assay Details
| Gene Symbol: LOC793439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109770.2 | NP_001103240.2 | 17 - 18 | 3622 | 68 | |
| Gene Symbol: tp53bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045153.1 | NP_001038618.1 | 16 - 17 | 3142 | 68 | |
| XM_005161809.4 | XP_005161866.1 | 17 - 18 | 3638 | 68 | ||
| XM_009296445.3 | XP_009294720.1 | 17 - 18 | 3635 | 68 | ||
More Information
Set Membership: |
|

LOC793439
Genomic Map