Assay Details
| Gene Symbol: LOC793439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109770.2 | NP_001103240.2 | 2 - 3 | 630 | 62 | |
| Gene Symbol: tp53bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045153.1 | NP_001038618.1 | 1 - 2 | 150 | 62 | |
| XM_005161809.4 | XP_005161866.1 | 2 - 3 | 646 | 62 | ||
| XM_009296445.3 | XP_009294720.1 | 2 - 3 | 646 | 62 | ||
| GenBank mRNA | BC142599.1 | - | 2 - 3 | 608 | 62 | |
More Information
Set Membership: |
|

LOC793439
Genomic Map