Assay Details
| Gene Symbol: tp53bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045153.1 | NP_001038618.1 | 5 - 6 | 613 | 70 | |
| XM_005161809.4 | XP_005161866.1 | 6 - 7 | 1109 | 70 | ||
| XM_009296445.3 | XP_009294720.1 | 6 - 7 | 1109 | 70 | ||
| GenBank mRNA | BC142599.1 | - | 6 - 7 | 1071 | 70 | |
| Gene Symbol: LOC793439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109770.2 | NP_001103240.2 | 6 - 7 | 1093 | 70 | |
More Information
Set Membership: |
|

tp53bp2b
Genomic Map