Assay Details
| Gene Symbol: serinc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045182.1 | NP_001038647.1 | 6 - 7 | 810 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

serinc2| Gene Symbol: serinc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045182.1 | NP_001038647.1 | 6 - 7 | 810 | 60 | |
Set Membership: |
|