Assay Details
| Gene Symbol: usp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045248.1 | NP_001038713.1 | 9 - 10 | 1587 | 62 | |
| XM_021480159.1 | XP_021335834.1 | 10 - 11 | 1667 | 62 | ||
| XM_021480160.1 | XP_021335835.1 | 10 - 11 | 1638 | 62 | ||
| GenBank mRNA | BC116508.1 | - | 9 - 10 | 1587 | 62 | |
| BC146618.1 | - | 10 - 11 | 1277 | 62 | ||
More Information
Set Membership: |
|

usp22
Genomic Map