Assay Details
| Gene Symbol: fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076569.1 | NP_001070037.1 | 6 - 7 | 689 | 111 | |
| XM_005162851.3 | XP_005162908.1 | 6 - 7 | 759 | 111 | ||
| XM_021470192.1 | XP_021325867.1 | 6 - 7 | 714 | 111 | ||
| XM_021470193.1 | XP_021325868.1 | 6 - 7 | 711 | 111 | ||
| GenBank mRNA | BC124506.1 | - | 6 - 7 | 689 | 111 | |
More Information
Set Membership: |
|

fam168b
Genomic Map