Assay Details
| Gene Symbol: b4galt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077259.2 | NP_001070727.2 | 1 - 2 | 670 | 99 | |
| GenBank mRNA | BC124813.1 | - | 1 - 2 | 550 | 99 | |
| FJ829869.1 | - | 1 - 2 | 272 | 99 | ||
More Information
Set Membership: |
|

b4galt1l
Genomic Map