Assay Details
| Gene Symbol: b4galt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC124813.1 | - | 2 - 3 | 784 | 72 | |
| FJ829869.1 | - | 2 - 3 | 506 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

b4galt1l| Gene Symbol: b4galt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC124813.1 | - | 2 - 3 | 784 | 72 | |
| FJ829869.1 | - | 2 - 3 | 506 | 72 | ||
Set Membership: |
|