Assay Details
| Gene Symbol: b4galt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077259.2 | NP_001070727.2 | 3 - 4 | 1094 | 76 | |
| XM_005157336.4 | XP_005157393.1 | 3 - 4 | 373 | 76 | ||
| GenBank mRNA | BC124813.1 | - | 3 - 4 | 974 | 76 | |
| FJ829869.1 | - | 3 - 4 | 696 | 76 | ||
More Information
Set Membership: |
|

b4galt1l
Genomic Map