Assay Details
| Gene Symbol: b4galt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077259.2 | NP_001070727.2 | 4 - 5 | 1211 | 97 | |
| XM_005157336.4 | XP_005157393.1 | 4 - 5 | 490 | 97 | ||
| GenBank mRNA | BC124813.1 | - | 4 - 5 | 1091 | 97 | |
| FJ829869.1 | - | 4 - 5 | 813 | 97 | ||
More Information
Set Membership: |
|

b4galt1l
Genomic Map