Assay Details
| Gene Symbol: ca15c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077333.1 | NP_001070801.1 | 2 - 3 | 261 | 103 | |
| XM_017358489.2 | XP_017213978.1 | 3 - 4 | 194 | 103 | ||
| GenBank mRNA | BC124780.1 | - | 2 - 3 | 261 | 103 | |
| BC164354.1 | - | 2 - 3 | 137 | 103 | ||
More Information
Set Membership: |
|

ca15c
Genomic Map