Assay Details
| Gene Symbol: ca15c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077333.1 | NP_001070801.1 | 6 - 7 | 760 | 89 | |
| XM_017358489.2 | XP_017213978.1 | 7 - 8 | 693 | 89 | ||
| GenBank mRNA | BC124780.1 | - | 6 - 7 | 760 | 89 | |
| BC164354.1 | - | 6 - 7 | 636 | 89 | ||
More Information
Set Membership: |
|

ca15c
Genomic Map