Assay Details
| Gene Symbol: dclk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077549.2 | NP_001071017.1 | 2 - 3 | 178 | 102 | |
| XM_005172671.4 | XP_005172728.1 | 6 - 7 | 1217 | 102 | ||
| XM_009305443.3 | XP_009303718.1 | 6 - 7 | 1244 | 102 | ||
| XM_009305445.3 | XP_009303720.1 | 6 - 7 | 1214 | 102 | ||
| GenBank mRNA | BC124721.1 | - | 2 - 3 | 178 | 102 | |
| BC165597.1 | - | 2 - 3 | 145 | 102 | ||
| EH432532.1 | - | 2 - 3 | 187 | 102 | ||
More Information
Set Membership: |
|

dclk1a
Genomic Map