Assay Details
| Gene Symbol: pcsk5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021476634.1 | XP_021332309.1 | 4 - 5 | 804 | 81 | |
| GenBank mRNA | DQ983232.1 | - | 4 - 5 | 733 | 81 | |
More Information
Set Membership: |
|

pcsk5a
Genomic Map