Assay Details
| Gene Symbol: asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080045.1 | NP_001073514.1 | 2 - 3 | 1280 | 76 | |
| XM_005167384.4 | XP_005167441.1 | 2 - 3 | 1373 | 76 | ||
| GenBank mRNA | BC125876.1 | - | 2 - 3 | 1280 | 76 | |
| KP324783.1 | - | 1 - 2 | 860 | 76 | ||
More Information
Set Membership: |
|

asphd2
Genomic Map