Assay Details
| Gene Symbol: git2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080188.2 | NP_001073657.2 | 14 - 15 | 1487 | 67 | |
| NM_001362634.1 | NP_001349563.1 | 14 - 15 | 1487 | 67 | ||
| XM_005155407.4 | XP_005155464.1 | 15 - 16 | 1579 | 67 | ||
| XM_005155409.4 | XP_005155466.1 | 15 - 16 | 1577 | 67 | ||
| GenBank mRNA | BC129227.1 | - | 14 - 15 | 1486 | 67 | |
More Information
Set Membership: |
|

git2b
Genomic Map