Assay Details
| Gene Symbol: arid6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080201.1 | NP_001073670.1 | 3 - 4 | 340 | 129 | |
| GenBank mRNA | BC129397.1 | - | 3 - 4 | 340 | 129 | |
More Information
Set Membership: |
|

arid6
Genomic Map