Assay Details
| Gene Symbol: g3bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080563.1 | NP_001074032.1 | 5 - 6 | 677 | 66 | |
| XM_021474934.1 | XP_021330609.1 | 5 - 6 | 1911 | 66 | ||
| GenBank mRNA | BC129213.1 | - | 5 - 6 | 677 | 66 | |
More Information
Set Membership: |
|

g3bp2
Genomic Map