Assay Details
| Gene Symbol: pax1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080592.1 | NP_001074061.1 | 2 - 3 | 865 | 104 | |
| GenBank mRNA | BC129448.1 | - | 2 - 3 | 865 | 104 | |
More Information
Set Membership: |
|

pax1a
Genomic Map