Assay Details
| Gene Symbol: pax1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080592.1 | NP_001074061.1 | 3 - 4 | 1072 | 91 | |
| GenBank mRNA | BC129448.1 | - | 3 - 4 | 1072 | 91 | |
More Information
Set Membership: |
|

pax1a
Genomic Map