Assay Details
| Gene Symbol: ccn1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080987.2 | NP_001074456.2 | 2 - 3 | 539 | 67 | |
| GenBank mRNA | BC085515.1 | - | 2 - 3 | 504 | 67 | |
| BC131858.1 | - | 2 - 3 | 464 | 67 | ||
| GQ273499.1 | - | 2 - 3 | 464 | 67 | ||
More Information
Set Membership: |
|

ccn1l2
Genomic Map