Assay Details
| Gene Symbol: ccn1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080987.2 | NP_001074456.2 | 3 - 4 | 850 | 103 | |
| GenBank mRNA | BC085515.1 | - | 3 - 4 | 815 | 103 | |
| BC131858.1 | - | 3 - 4 | 775 | 103 | ||
| GQ273499.1 | - | 3 - 4 | 775 | 103 | ||
More Information
Set Membership: |
|

ccn1l2
Genomic Map