Assay Details
| Gene Symbol: rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082856.1 | NP_001076325.1 | 3 - 4 | 654 | 73 | |
| GenBank mRNA | BC096929.1 | - | 4 - 5 | 847 | 73 | |
| DQ017626.1 | - | 3 - 4 | 618 | 73 | ||
| EF107093.1 | - | 3 - 4 | 654 | 73 | ||
More Information
Set Membership: |
|

rorb
Genomic Map