Assay Details
| Gene Symbol: itgb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082948.1 | NP_001076417.1 | 10 - 11 | 1771 | 76 | |
| GenBank mRNA | BC163207.1 | - | 10 - 11 | 1730 | 76 | |
| DQ178646.1 | - | 10 - 11 | 1771 | 76 | ||
More Information
Set Membership: |
|

itgb3b
Genomic Map