Assay Details
| Gene Symbol: itgb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082948.1 | NP_001076417.1 | 6 - 7 | 1022 | 117 | |
| GenBank mRNA | BC163207.1 | - | 6 - 7 | 981 | 117 | |
| DQ178646.1 | - | 6 - 7 | 1022 | 117 | ||
More Information
Set Membership: |
|

itgb3b
Genomic Map