Assay Details
| Gene Symbol: stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089448.1 | NP_001082917.1 | 9 - 10 | 1035 | 118 | |
| XM_005159799.4 | XP_005159856.1 | 10 - 11 | 1057 | 118 | ||
| GenBank mRNA | BC134162.1 | - | 9 - 10 | 1035 | 118 | |
More Information
Set Membership: |
|

stra6l
Genomic Map