Assay Details
| Gene Symbol: ube2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089453.1 | NP_001082922.1 | 5 - 6 | 482 | 64 | |
| GenBank mRNA | BC139559.1 | - | 5 - 6 | 482 | 64 | |
More Information
Set Membership: |
|

ube2d4
Genomic Map