Assay Details
| Gene Symbol: cdx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098762.1 | NP_001092232.1 | 2 - 3 | 614 | 63 | |
| GenBank mRNA | AY761094.1 | - | 2 - 3 | 1166 | 63 | |
| BC142877.1 | - | 2 - 3 | 614 | 63 | ||
More Information
Set Membership: |
|

cdx1b
Genomic Map