Assay Details
| Gene Symbol: tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099263.1 | NP_001092733.1 | 6 - 7 | 605 | 69 | |
| GenBank mRNA | BC142903.1 | - | 6 - 7 | 605 | 69 | |
More Information
Set Membership: |
|

tm2d1
Genomic Map